Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGAGGTTATTATAACCTATTACT[A/G]AAAGGATGTTTGCATAACAGCACAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ndst4 PubMed Links |
Ndst4 - N-deacetylase/N-sulfotransferase (heparin glucosaminyl) 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022565.2 | Intron | NP_072087.2 |