Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACTCACAGCTGGCTGCTTTCAAC[A/C]TTTACGGTAGATTGTAGGAAGCTGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ank1 PubMed Links |
Ank1 - ankyrin 1, erythroid | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110783.3 | Intron | NP_001104253.1 | ||||
NM_001277280.2 | Intron | NP_001264209.1 | ||||
NM_001277281.2 | Intron | NP_001264210.1 | ||||
NM_001277284.2 | Intron | NP_001264213.1 | ||||
NM_001277286.2 | Intron | NP_001264215.1 | ||||
NM_001277289.2 | Intron | NP_001264218.1 | ||||
NM_031158.4 | Intron | NP_112435.2 |