Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCACAGGAGAAAAGTCCTTACG[A/T]GGTTCCCTAAAGTAGAGGGGATTTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700011M02Rik PubMed Links |
1700011M02Rik - RIKEN cDNA 1700011M02 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Dmrtc1a - DMRT-like family C1a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038616.2 | 81 | Intron | NP_001033705.2 | |||
NM_027591.3 | 81 | Silent Mutation | NP_081867.1 | |||
NM_029378.2 | 81 | Intron | NP_083654.1 |