Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACAGAAGATGATAGAAAATAAAG[C/G]AGAGATTATGGAAGTAACCCACAGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Adamts19 PubMed Links |
Adamts19 - a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175506.3 | Intron | NP_780715.1 |