Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGTGATCATGGTTTTACAAGGC[C/G]TTGGGGCAGATCTTTAAGTCACCAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Frem1 PubMed Links |
Frem1 - Fras1 related extracellular matrix protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198811.1 | Intron | NP_001185740.1 | ||||
NM_177863.4 | Intron | NP_808531.2 |