Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTACCTTCCTAGCCCCCCGCAC[A/G]GTTCCGTGAGTGGCTGAGTGATCAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Zdhhc21 PubMed Links |
Zdhhc21 - zinc finger, DHHC domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026647.3 | 7640 | UTR 3 | NP_080923.2 |