Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAGAGTCCCATGTTAGTGTGAT[A/G]CTTGTAGCTGTGCTCTCGGTCACTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ptprd PubMed Links |
Ptprd - protein tyrosine phosphatase, receptor type, D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011211.3 | 1914 | Silent Mutation | NP_035341.2 |