Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATAGCCAGATTCTTCACTGTAAA[C/T]GGGCACTTTTTGAAGTACACTCTCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Epha3 PubMed Links |
Epha3 - Eph receptor A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010140.3 | 719 | Silent Mutation | NP_034270.1 |