Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTCTGATAACAGCTTTATTGCA[A/G]TGTGCCATGCACATTCTTGTATTTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rb1cc1 PubMed Links |
Rb1cc1 - RB1-inducible coiled-coil 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009826.4 | Intron | NP_033956.2 |