Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAAATCACAGCCACTGAGCCTCAG[T/C]GAACTGAGTGACCTTTCCACAGCCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Creb5 PubMed Links |
Creb5 - cAMP responsive element binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172728.2 | Intron | NP_766316.1 |