Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAACACAAGATCATAGTTAACTG[A/G]AAACTACATCTTTCCTCCCAGTTGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sncaip PubMed Links |
Sncaip - synuclein, alpha interacting protein (synphilin) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199151.1 | Intron | NP_001186080.1 | ||||
NM_001199153.1 | Intron | NP_001186082.1 | ||||
NM_001199154.1 | Intron | NP_001186083.1 | ||||
NM_026408.4 | Intron | NP_080684.3 |