Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGCTTTATTAGCGCTGCTTGAC[A/C]CTGATGGACAGAGGATTTGTTTTCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dock11 PubMed Links |
Dock11 - dedicator of cytokinesis 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009947.3 | 3207 | Silent Mutation | NP_001009947.2 |