Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTATCTCTAGACTAGAGACTCC[A/G]TTTCTCGTTTCTCTTCTCTGTTAAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Enox2 PubMed Links |
Enox2 - ecto-NOX disulfide-thiol exchanger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271447.1 | Intron | NP_001258376.1 | ||||
NM_001271448.1 | Intron | NP_001258377.1 | ||||
NM_001271449.1 | Intron | NP_001258378.1 | ||||
NM_001271450.1 | Intron | NP_001258379.1 | ||||
NM_001271451.1 | Intron | NP_001258380.1 | ||||
NM_145951.5 | Intron | NP_666063.1 |