Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATTATTTTTTATCCATAATTTT[A/C]CTGCTATTCATGATTACCATAATTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cntnap2 PubMed Links |
Cntnap2 - contactin associated protein-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004357.2 | Intron | NP_001004357.2 | ||||
NM_025771.3 | Intron | NP_080047.1 |