Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTATAACTTACCCATTATGTTCT[G/T]ATAAAATTCTATCTGAGCCCAGAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
D030040B21Rik PubMed Links |
D030040B21Rik - RIKEN cDNA D030040B21 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Tceb1 - transcription elongation factor B (SIII), polypeptide 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026456.4 | 85 | UTR 5 | NP_080732.1 |