Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACACTACTTGCTCTACTACAGAT[G/T]TTGATGGATATCTGTGATAGTAGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kdm4c PubMed Links |
Kdm4c - lysine (K)-specific demethylase 4C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172095.1 | Intron | NP_001165566.1 | ||||
NM_144787.2 | Intron | NP_659036.1 |