Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTGGCGCTGGAGCTGGTTATCGC[C/T]GCGCTGGCGGTGGCGGGCAACGTGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Adora2b PubMed Links |
Adora2b - adenosine A2b receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007413.4 | 172 | Silent Mutation | NP_031439.2 |