Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAGAAGATATGCTCTAATCCTC[C/T]TTCCTAAGGTTGCTCATTTAAAAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dnah9 PubMed Links |
Dnah9 - dynein, axonemal, heavy chain 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099633.1 | Intron | NP_001093103.1 |