Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCCGGGGGCTGATGAAGCGCAT[C/T]TCCTTCAGAAGTTCCTCCTTCTCAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Wwc1 PubMed Links |
Wwc1 - WW, C2 and coiled-coil domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_170779.1 | 1059 | Silent Mutation | NP_740749.1 |