Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gucy1a3 PubMed Links |
Gucy1a3 - guanylate cyclase 1, soluble, alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021896.5 | 617 | Missense Mutation | NP_068696.2 |
Mir7010 - microRNA 7010 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |