Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCATAGTGTGTGATGTTAGGGGA[C/T]GCCTCAATGTGTAGCATGATGTATT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Greb1 PubMed Links |
Greb1 - gene regulated by estrogen in breast cancer protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252071.1 | 5697 | Silent Mutation | NP_001239000.1 | |||
NM_015764.4 | 5697 | Silent Mutation | NP_056579.2 |