Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGAAGGAAGCACTTGGTTAATG[C/T]ATTGTTCTTGATTTATGGCACACTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Astn2 PubMed Links |
Astn2 - astrotactin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019514.3 | Intron | NP_062387.2 | ||||
NM_207109.2 | Intron | NP_996992.1 |