Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTCCAAGTATGAGCACAGTTAC[C/T]GGGTTCCAGCAGACCCAGTTACAGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hif1a PubMed Links |
Hif1a - hypoxia inducible factor 1, alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010431.2 | 2253 | Silent Mutation | NP_034561.2 |