Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Il11 PubMed Links |
Il11 - interleukin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008350.4 | 19 | Intron | NP_032376.1 |
Rpl28 - ribosomal protein L28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Tmem190 - transmembrane protein 190 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030028.1 | 19 | UTR 5 | NP_084304.1 |
Tmem238 - transmembrane protein 238 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |