Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGTCCACGTAGACCCCCTAATGG[C/G]CATACAGGTGCTGAATCTATGTAAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kcnmb2 PubMed Links |
Kcnmb2 - potassium large conductance calcium-activated channel, subfamily M, beta member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_028231.2 | Intron | NP_082507.1 |