Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCATTTGGTTTGTATTGATTTGA[C/T]CAACACTATCTACGCATGTCCCATG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Akap6 PubMed Links |
Akap6 - A kinase (PRKA) anchor protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198111.2 | 9780 | UTR 3 | NP_932779.2 |