Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTATAACAATAACTTACAAATAC[G/T]GGGAAAGGAAGCTGGGAAAGAACGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Phlpp1 PubMed Links |
Phlpp1 - PH domain and leucine rich repeat protein phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133821.3 | Intron | NP_598582.3 |