Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAATCCAACCTGATTGCCTAACT[G/T]GAAGTACTTTCATGGAGACGGGGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rnf145 PubMed Links |
Rnf145 - ring finger protein 145 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166553.1 | Intron | NP_001160025.1 | ||||
NM_028862.3 | Intron | NP_083138.2 |