Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACTGGACCGGGCCTGAATTGCTG[A/G]CTCTTGTGGTGACAGAGTGTAATAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ccnjl PubMed Links |
Ccnjl - cyclin J-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001045530.2 | Intron | NP_001038995.1 |