Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ccdc125 PubMed Links |
Ccdc125 - coiled-coil domain containing 125 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168386.2 | 1783 | UTR 3 | NP_001161858.1 | |||
NM_183115.5 | 1783 | Missense Mutation | NP_898938.4 |
Cdk7 - cyclin-dependent kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009874.3 | 1783 | Intron | NP_034004.2 |