Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Adam17 PubMed Links |
Adam17 - a disintegrin and metallopeptidase domain 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277266.1 | Intron | NP_001264195.1 | ||||
NM_009615.6 | Intron | NP_033745.4 |
Iah1 - isoamyl acetate-hydrolyzing esterase 1 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |