Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAACAAGACAATCTCAAGTGACC[A/G]CTATTTTTGTGAATCATAAAATTCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Bod1l PubMed Links |
Bod1l - biorientation of chromosomes in cell division 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081422.2 | Intron | NP_001074891.1 |