Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTTGCTGTTATGACTTGACAGT[A/G]CTGCACTTCTGTTATGAATTCCTAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Polk PubMed Links |
Polk - polymerase (DNA directed), kappa | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012048.2 | Intron | NP_036178.1 |