Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATATTCTATGGAGGGGCTGAAAA[C/T]TCCTCCATAGGCTGTGCTTTGAAAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hdac4 PubMed Links |
Hdac4 - histone deacetylase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207225.1 | Intron | NP_997108.1 |