Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCCAGCAAAGGATTGTGTAGGAA[C/T]TCACTTTTCTTCATATCCTCACCAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Edil3 PubMed Links |
Edil3 - EGF-like repeats and discoidin I-like domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037987.3 | Intron | NP_001033076.1 | ||||
NM_010103.4 | Intron | NP_034233.1 |