Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gtpbp6 PubMed Links |
Gtpbp6 - GTP binding protein 6 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145147.5 | 2771 | Missense Mutation | NP_660129.2 |
Plcxd1 - phosphatidylinositol-specific phospholipase C, X domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281812.1 | 2771 | UTR 3 | NP_001268741.1 | |||
NM_207279.2 | 2771 | UTR 3 | NP_997162.2 |
Zfp605 - zinc finger protein 605 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |