Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTGCCTTACCAGGGTTTGGTGC[G/T]CAAACTACACAACTTTCAGGCTTGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Abca13 PubMed Links |
Abca13 - ATP-binding cassette, sub-family A (ABC1), member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178259.3 | 5503 | Missense Mutation | NP_839990.2 |