Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGATGTACACAGTCTAAGCGCG[C/G]CGAGACAAGGAAACTCAGACGCTCG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pik3cg PubMed Links |
Pik3cg - phosphoinositide-3-kinase, catalytic, gamma polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146200.1 | 97 | Intron | NP_001139672.1 | |||
NM_001146201.1 | 97 | Intron | NP_001139673.1 | |||
NM_020272.2 | 97 | UTR 5 | NP_064668.2 |