Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCTGCCAGGTGGACAAGGGATT[C/T]CCAGACGCTGCAGCTCCACGCGGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cdkl4 PubMed Links |
Cdkl4 - cyclin-dependent kinase-like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033443.4 | Intron | NP_001028615.1 |
F420015M19Rik - RIKEN cDNA F420015M19 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |