Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAGTTGCTATGGTGTTTTTTTTT[C/T]CCTCCCCCTTAGCTACTGATTTTTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tmem178 PubMed Links |
Tmem178 - transmembrane protein 178 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026516.2 | Intron | NP_080792.2 |