Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGTGCCCAAGCCACATAGCAGG[C/T]ACCTCTGGGTCCTCGCTCTCCTTCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Inhba PubMed Links |
Inhba - inhibin beta-A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008380.1 | 1246 | Silent Mutation | NP_032406.1 |