Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGAATGCTGAAATTTAGAAAA[C/T]GTAGTCTTAACCCTGGGTGACTAGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Plcb1 PubMed Links |
Plcb1 - phospholipase C, beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145830.1 | Intron | NP_001139302.1 | ||||
NM_019677.2 | Intron | NP_062651.2 |