Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCGCTGCCCTTCTCGGTGCCC[A/G]CCCAAGTGCCCCAGTATATCACCGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nov PubMed Links |
Nov - nephroblastoma overexpressed gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010930.4 | 146 | Silent Mutation | NP_035060.1 |