Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAATAGCCTCCCTAGCACCATT[G/T]GGTGGTAAAGGAGAGAGTAAAATAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pard3b PubMed Links |
Pard3b - par-3 family cell polarity regulator beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081050.2 | Intron | NP_001074519.2 |