Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACATGCTACCCTCTTCAAAGAAT[A/G]CAAACAAACAGCTGTCATTTTGGAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kcnq3 PubMed Links |
Kcnq3 - potassium voltage-gated channel, subfamily Q, member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152923.2 | Intron | NP_690887.2 |