Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACCAGTTTCACAGTTAGCAAGTA[G/T]GAAGGCTGGATGCCTGTGGTTCCTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ranbp17 PubMed Links |
Ranbp17 - RAN binding protein 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023146.2 | Intron | NP_075635.1 |