Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTCATCATTTGGATTTCAGATT[C/T]GGAAATGGCTTTCATGCCGTTATTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nrxn1 PubMed Links |
Nrxn1 - neurexin I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020252.3 | Intron | NP_064648.3 | ||||
NM_177284.2 | Intron | NP_796258.2 |