Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGAGGGTCATACAAAATGTGTG[C/G]AACTTTTGCTGTCCAGTGGGGCGGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Asb3 PubMed Links |
Asb3 - ankyrin repeat and SOCS box-containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023906.3 | 870 | Missense Mutation | NP_076395.2 |