Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAACCCTAGTGCACCCACATGCT[C/T]CCCTCAAGCTGAGTGACAGAGTCTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slc45a4 PubMed Links |
Slc45a4 - solute carrier family 45, member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033219.3 | Intron | NP_001028391.2 | ||||
NM_001168255.1 | Intron | NP_001161727.1 |