Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGCGTAAAAGGCGCACCACCTT[C/T]AAAGAGCTGAAGAGACTGCTGATTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kcnh5 PubMed Links |
Kcnh5 - potassium voltage-gated channel, subfamily H (eag-related), member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172805.3 | 1773 | Silent Mutation | NP_766393.2 |